Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014240.3 | NP_001014262.3 | 2 - 3 | 282 | 162 | |
GenBank mRNA | AY327508.1 | - | 2 - 3 | 318 | 162 | |
BC088169.1 | - | 2 - 3 | 282 | 7322153 | 162 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |