Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030020.1 | NP_001025191.1 | 11 | 112 | ||
GenBank mRNA | BI298837.1 | - | 1 | 372 | 112 | |
CX569141.1 | - | 1 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |