Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000117.1 | NP_001000117.1 | 1 | 67 | 106 |
Gene Symbol: LOC100910993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752171.3 | XP_003752219.1 | 1 | 136 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |