Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199083.2 | NP_954514.2 | 5 - 6 | 652 | 59 | |
GenBank mRNA | AY344057.1 | - | 5 - 6 | 652 | 59 | |
BC088091.1 | - | 5 - 6 | 629 | 7325613 | 59 | |
FQ215167.1 | - | 5 - 6 | 619 | 59 | ||
FQ232331.1 | - | 5 - 6 | 632 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |