Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199119.1 | NP_954550.1 | 1 - 2 | 48 | 60 | |
GenBank mRNA | AY341876.1 | - | 1 - 2 | 48 | 60 | |
BC097262.1 | - | 1 - 2 | 35 | 7442976 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |