Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203337.2 | NP_976082.1 | 7 - 8 | 671 | 72 | |
XM_006242789.2 | XP_006242851.1 | 8 - 9 | 787 | 72 | ||
XM_008766058.1 | XP_008764280.1 | 7 - 8 | 1173 | 72 | ||
XM_008766059.1 | XP_008764281.1 | 7 - 8 | 1162 | 72 | ||
XM_008766060.1 | XP_008764282.1 | 7 - 8 | 1621 | 72 | ||
GenBank mRNA | AJ626825.1 | - | 6 - 7 | 435 | 72 | |
BC089057.1 | - | 7 - 8 | 671 | 7315905 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |