Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044293.1 | NP_001037758.1 | 2 | 1293 | 128 | |
XM_006236587.2 | XP_006236649.1 | 2 | 2697 | 128 | ||
GenBank mRNA | BC087702.1 | - | 2 | 1293 | 7313964 | 128 |
FQ212618.1 | - | 2 | 1287 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |