Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025563.1 | NP_001020734.1 | 6 | 1674 | 81 | |
XM_006249547.2 | XP_006249609.1 | 6 | 1811 | 81 | ||
XM_008769330.1 | XP_008767552.1 | 6 | 1819 | 81 | ||
XM_008769331.1 | XP_008767553.1 | 6 | 1815 | 81 | ||
GenBank mRNA | BC161821.1 | - | 6 | 1634 | 9027947 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |