Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751162.3 | XP_003751210.1 | 7 | 4158 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |