Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022697.2 | NP_073188.2 | 4 | 1491 | 141 | |
GenBank mRNA | BC081800.1 | - | 4 | 1491 | 7131175 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |