Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053309.1 | NP_445761.1 | 2 | 399 | 291 | |
GenBank mRNA | AB007689.1 | - | 2 | 399 | 291 | |
AB007690.1 | - | 2 | 213 | 291 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Amplicon spans exons and probe does not span exons |