Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244171.2 | XP_006244233.1 | 13 | 2544 | 178 | |
XM_236774.7 | XP_236774.5 | 13 | 2522 | 178 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |