Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044227.1 | NP_001037692.1 | 2 - 3 | 302 | 69 | |
XM_006249941.1 | XP_006250003.1 | 3 - 4 | 413 | 69 | ||
GenBank mRNA | BC088173.1 | - | 2 - 3 | 302 | 7323337 | 69 |
FQ218705.1 | - | 2 - 3 | 320 | 69 | ||
FQ219461.1 | - | 2 - 3 | 323 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |