Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008752.1 | NP_001008752.1 | 2 - 3 | 583 | 142 | |
XM_006247309.2 | XP_006247371.1 | 2 - 3 | 661 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |