Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ011608.1 | - | 6 - 7 | 643 | 127 | |
BC088272.1 | - | 6 - 7 | 650 | 7379940 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |