Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008899.1 | NP_001008899.1 | 4 - 5 | 1015 | 165 | |
XM_006242436.2 | XP_006242498.1 | 4 - 5 | 1519 | 165 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |