Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009268.1 | NP_001009268.1 | 2 - 3 | 221 | 131 | |
GenBank mRNA | BC088442.1 | - | 2 - 3 | 221 | 7126325 | 131 |
FQ212547.1 | - | 2 - 3 | 220 | 131 | ||
FQ219071.1 | - | 2 - 3 | 170 | 131 | ||
FQ219838.1 | - | 2 - 3 | 187 | 131 | ||
FQ225424.1 | - | 2 - 3 | 192 | 131 | ||
FQ226083.1 | - | 2 - 3 | 131 | |||
FQ226422.1 | - | 2 - 3 | 170 | 131 | ||
FQ227142.1 | - | 2 - 3 | 170 | 131 | ||
FQ227488.1 | - | 2 - 3 | 219 | 131 | ||
FQ227830.1 | - | 2 - 3 | 170 | 131 | ||
FQ230874.1 | - | 2 - 3 | 205 | 131 | ||
FQ231828.1 | - | 2 - 3 | 172 | 131 | ||
FQ231874.1 | - | 2 - 3 | 205 | 131 | ||
FQ234230.1 | - | 2 - 3 | 172 | 131 | ||
FQ235071.1 | - | 2 - 3 | 192 | 131 | ||
FQ235201.1 | - | 2 - 3 | 187 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |