Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009504.1 | NP_001009504.1 | 2 - 3 | 1140 | 113 | |
XM_006246020.2 | XP_006246082.1 | 3 - 4 | 1458 | 113 | ||
GenBank mRNA | AY823669.1 | - | 2 - 3 | 1140 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |