Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009524.1 | NP_001009524.1 | 15 - 16 | 1693 | 150 | |
XM_006242737.2 | XP_006242799.1 | 15 - 16 | 150 | |||
GenBank mRNA | AF299298.1 | - | 15 - 16 | 150 | ||
AF414431.1 | - | 15 - 16 | 1547 | 150 | ||
AY040870.1 | - | 15 - 16 | 1693 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |