Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009524.1 | NP_001009524.1 | 4 - 5 | 614 | 171 | |
XM_006242737.2 | XP_006242799.1 | 4 - 5 | 492 | 171 | ||
GenBank mRNA | AF299297.1 | - | 4 - 5 | 614 | 171 | |
AF299298.1 | - | 4 - 5 | 468 | 171 | ||
AF414431.1 | - | 4 - 5 | 468 | 171 | ||
AY040870.1 | - | 4 - 5 | 614 | 171 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |