Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5sl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009705.1 | NP_001009705.1 | 3 - 4 | 304 | 154 | |
XM_006228449.2 | XP_006228511.1 | 3 - 4 | 431 | 154 | ||
GenBank mRNA | BC088283.1 | - | 3 - 4 | 304 | 7384054 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |