Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009825.1 | NP_001009825.1 | 6 - 7 | 606 | 131 | |
XM_008767362.1 | XP_008765584.1 | 6 - 7 | 1552 | 131 | ||
GenBank mRNA | BC088868.1 | - | 6 - 7 | 606 | 7321115 | 131 |
FQ233576.1 | - | 620 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |