Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053570.1 | NP_446022.1 | 5 - 6 | 771 | 169 | |
XM_006248000.2 | XP_006248062.1 | 5 - 6 | 872 | 169 | ||
GenBank mRNA | AF109643.1 | - | 5 - 6 | 700 | 169 | |
AF109644.1 | - | 5 - 6 | 700 | 169 | ||
BC088313.1 | - | 5 - 6 | 771 | 7324723 | 169 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |