Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009492.1 | NP_001009492.1 | 4 - 5 | 930 | 107 | |
XM_008769248.1 | XP_008767470.1 | 4 - 5 | 931 | 107 | ||
GenBank mRNA | AY196698.1 | - | 4 - 5 | 930 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |