Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49i7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009500.1 | NP_001009500.1 | 3 - 4 | 423 | 149 | |
XM_008763320.1 | XP_008761542.1 | 4 - 5 | 1520 | 149 | ||
GenBank mRNA | AY659934.1 | - | 3 - 4 | 423 | 149 |
Gene Symbol: LOC100911272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763440.1 | XP_008761662.1 | 4 - 5 | 1279 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |