Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012000.1 | NP_001012000.1 | 3 - 4 | 461 | 142 | |
GenBank mRNA | AY548832.1 | - | 3 - 4 | 461 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |