Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012233.1 | NP_001012233.1 | 2 - 3 | 144 | 159 | |
GenBank mRNA | AB164465.1 | - | 2 - 3 | 144 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |