Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022386.2 | NP_071781.1 | 2 - 3 | 225 | 154 | |
XM_006247913.2 | XP_006247975.1 | 3 - 4 | 3309 | 154 | ||
XM_006247921.2 | XP_006247983.1 | 2 - 3 | 986 | 154 | ||
XM_006247922.2 | XP_006247984.1 | 2 - 3 | 147 | 154 | ||
XM_006247923.2 | XP_006247985.1 | 2 - 3 | 916 | 154 | ||
XM_008768482.1 | XP_008766704.1 | 2 - 3 | 3758 | 154 | ||
GenBank mRNA | AB026487.1 | - | 1 - 2 | 99 | 154 | |
BC078828.1 | - | 2 - 3 | 225 | 7127046 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |