Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008808.1 | NP_001008808.1 | 2 - 3 | 654 | 125 | |
XM_008765703.1 | XP_008763925.1 | 2 - 3 | 694 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |