Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008823.1 | NP_001008823.1 | 5 - 6 | 920 | 71 | |
XM_006247390.2 | XP_006247452.1 | 5 - 6 | 1508 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |