Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008849.1 | NP_001008849.1 | 3 - 4 | 615 | 145 | |
XM_006255884.2 | XP_006255946.1 | 4 - 5 | 1037 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |