Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012748.1 | NP_001012766.1 | 6 - 7 | 1263 | 102 | |
GenBank mRNA | AB121033.1 | - | 6 - 7 | 1263 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |