Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024286.1 | NP_001019457.1 | 2 - 3 | 64 | 66 | |
XM_008759404.1 | XP_008757626.1 | 1 - 2 | 761 | 66 | ||
GenBank mRNA | BC089881.1 | - | 2 - 3 | 64 | 7368473 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |