Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024742.1 | NP_001019913.1 | 1 - 2 | 828 | 101 | |
XM_006250697.2 | XP_006250759.1 | 2 - 4 | 810 | 101 | ||
GenBank mRNA | BC078912.1 | - | 1 - 2 | 801 | 101 | |
BC097350.1 | - | 1 - 2 | 828 | 7462336 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |