Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025113.1 | NP_001020284.1 | 4 - 5 | 423 | 132 | |
XM_006256442.2 | XP_006256504.1 | 5 - 6 | 603 | 132 | ||
XM_008772879.1 | XP_008771101.1 | 3 - 4 | 278 | 132 | ||
GenBank mRNA | AY779029.1 | - | 4 - 5 | 423 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |