Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033914.1 | NP_001029086.1 | 7 - 8 | 1587 | 126 | |
NM_021852.2 | NP_068624.2 | 6 - 7 | 1416 | 126 | ||
XM_006246521.2 | XP_006246583.1 | 7 - 8 | 1371 | 126 | ||
XM_006246522.2 | XP_006246584.1 | 8 - 9 | 1669 | 126 | ||
XM_006246523.2 | XP_006246585.1 | 6 - 7 | 1200 | 126 | ||
GenBank mRNA | AF096269.1 | - | 5 - 6 | 1157 | 126 | |
BC094524.1 | - | 7 - 8 | 1566 | 7318094 | 126 | |
FQ211574.1 | - | 8 - 9 | 1669 | 126 | ||
FQ211939.1 | - | 7 - 8 | 1611 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |