Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037360.1 | NP_001032437.1 | 1 | 1061 | 94 | |
GenBank mRNA | BC104717.1 | - | 1 | 1061 | 7381762 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |