Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047095.1 | NP_001040560.1 | 4 - 5 | 456 | 147 | |
GenBank mRNA | AB256043.1 | - | 4 - 5 | 456 | 147 | |
AM258958.1 | - | 4 - 5 | 456 | 147 | ||
BC158562.1 | - | 4 - 5 | 460 | 9088902 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |