Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077826.1 | NP_001071294.1 | 4 - 5 | 548 | 83 | |
GenBank mRNA | BC158557.1 | - | 651 | 7313337 | 83 | |
EF044301.1 | - | 4 - 5 | 548 | 83 | ||
FQ213412.1 | - | 4 - 5 | 559 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |