Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008966.1 | NP_001008966.1 | 1 | 566 | 113 | |
GenBank mRNA | AY510353.1 | - | 1 | 566 | 113 |
Gene Symbol: LOC103691019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758872.1 | XP_008757094.1 | 1 | 566 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |