Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077590.1 | NP_001071058.1 | 7 | 1420 | 121 | |
GenBank mRNA | AY786529.2 | - | 7 | 1420 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |