Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008870.2 | NP_001008870.1 | 1 - 2 | 42 | 70 |
Gene Symbol: LOC100910138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751350.3 | XP_003751398.1 | 1 - 2 | 147 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |