Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 8 - 9 | 677 | 61 | |
GenBank mRNA | CA512184.1 | - | 3 - 4 | 301 | 61 | |
FQ223839.1 | - | 315 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |