Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045843.1 | NP_001039308.1 | 1 | 1549 | 95 | |
GenBank mRNA | BC098703.1 | - | 1 | 1549 | 7389453 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |