Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107712.2 | NP_001101182.2 | 4 - 5 | 476 | 90 | |
XM_006233087.1 | XP_006233149.1 | 3 - 4 | 453 | 90 | ||
GenBank mRNA | CB724536.1 | - | 4 - 5 | 494 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |