Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Imp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108152.1 | NP_001101622.1 | 1 | 250 | 103 | |
GenBank mRNA | BC166985.1 | - | 256 | 7322293 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |