Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114621.1 | NP_001108093.1 | 16 - 17 | 2221 | 80 | |
XM_008762082.1 | XP_008760304.1 | 18 - 19 | 3621 | 80 | ||
XM_008762083.1 | XP_008760305.1 | 12 - 13 | 2353 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |