Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spetex-2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009969.1 | NP_001009969.1 | 1 | 49 | 104 | |
GenBank mRNA | AB180083.1 | - | 1 | 49 | 104 |
Gene Symbol: Spetex-2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011697.1 | NP_001011697.1 | 1 | 104 | ||
GenBank mRNA | AB180077.1 | - | 1 | 104 |
Gene Symbol: Spetex-2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011698.1 | NP_001011698.1 | 1 | 49 | 104 | |
GenBank mRNA | AB180078.1 | - | 1 | 49 | 104 |
Gene Symbol: Spetex-2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009968.1 | NP_001009968.1 | 1 | 49 | 104 | |
GenBank mRNA | AB180081.1 | - | 1 | 49 | 104 |
Gene Symbol: Spetex-2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009971.1 | NP_001009971.1 | 1 | 49 | 104 | |
GenBank mRNA | AB180082.1 | - | 1 | 49 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Amplicon spans exons and probe does not span exons |