Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099457.1 | NP_001092927.1 | 3 | 1046 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Rattus norvegicus Amplicon spans exons and probe does not span exons |