Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100681.1 | NP_001094151.1 | 1 - 2 | 452 | 71 | |
NM_001134862.1 | NP_001128334.1 | 1 - 2 | 622 | 71 | ||
GenBank mRNA | BC161983.1 | - | 1 - 2 | 452 | 9030389 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |